About this item
Rat monoclonal IgG2b antibody against mouse CD25, FITC-conjugated.
- T cells
- T cells Regulatory
The 3C7 antibody reacts with murine CD25 (interleukin‐2 receptor α chain; IL‐2Rα), an ~55 kDa type 1 transmembrane glycoprotein expressed on the surface of T and B cell progenitors and activated mature B and T cells. CD25 has been used as a marker to identify CD4+/FoxP3+ regulatory T cells in mice. Mutations in CD25 result in severe immunodeficiency, underscoring the importance of the roles the protein plays in the differentiation, activation and proliferation of lymphocytes, and in the maintenance of self-tolerance. CD25 per se has low affinity for its IL-2 ligand but forms dimers with CD122 (IL-2Rβ) and CD132 (IL-2Rγ) that together associate to form the IL-2R receptor, which binds IL-2 with high affinity. CD25 acts to increase the specificity and affinity of IL-2 binding and is necessary for receptor clustering and signal transduction by the complex. Binding of the 3C7 antibody inhibits binding of IL‐2 to free CD25 and the IL-2R receptor. The epitope is distinct from those recognised by the anti-CD25 antibody clones 7D4 and PC61. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse T Cell Enrichment Kit (Catalog #19751).
Type:
Antigen: CD25
Clonality: Monoclonal
Clone: 3C7
Conjugation: FITC
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse
Specifications
- Catalog No:
- STMC60010FI
- STMC60010FI.1
- Antigen name:
- CD25
- CD25
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- 3C7
- 3C7
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 16184
- 16184
- Isotype:
- IgG2b kappa
- IgG2b kappa