Order Entry
Slovenia
ContactUsLinkComponent
 
Anti-CD45RA Hamster Monoclonal Antibody [clone: HI100] (Biotinilated)
 
undefined
Anti-CD45RA Hamster Monoclonal Antibody [clone: HI100] (Biotinilated)

 

  • Catalog No:
  • STMC100-0322
  • STMC100-0321
  • Antigen name:
  • CD45RA
  • CD45Ra
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • HI100
  • HI100
  • Reactivity:
  • Chimpanzee, Human
  • Chimpanzee, Human
  • Host:
  • Hamster
  • Hamster
  • Gene ID:
  • 5788
  • 5788

 

Frequently Bought Together