Order Entry
Slovenia
ContactUsLinkComponent
Super Nuova+™ Hotplate Stirrer
Super Nuova+™ Hotplate Stirrer
Certificates
undefined
Super Nuova+™ Hotplate Stirrer

Specifications

  • Catalog No:
  • BARNSP88857198
  • BARNSP88857195
  • BARNSP88850195
  • Plate material:
  • Aluminium
  • Ceramic
  • Ceramic
  • Electrical:
  • 230 V, 50/60 Hz
  • 230 V, 50/60 Hz
  • 230 V, 50/60 Hz
  • Speed range:
  • 50 - 1500 rpm
  • 50 - 1500 rpm
  • 50 - 1500 rpm
  • W×D×H:
  • 208×330×97 mm
  • 208×330×97 mm
  • 287×411×102 mm
  • Temperature range:
  • 300 °C
  • 450 °C
  • 400 °C
  • Weight:
  • 5,5 kg
  • 5,6 kg
  • 8 kg

Specifications

Frequently Bought Together