Order Entry
Slovenia
ContactUsLinkComponent
 
Acridine Orange 10-nonyl bromide (NAO), Sigma-Aldrich®
Catalog # A7847-100MG
CAS Number:  
undefined
Acridine Orange 10-nonyl bromide (NAO), Sigma-Aldrich®
Catalog # A7847-100MG
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance (Colour):
    Orange to Red
  • Appearance (Form):
    Powder or Solid
  • Proton NMR Spectrum:
    Conforms to Structure
  • EmM at lambda max 491 to 497 nm in Methanol:
    82 - 94
  • Wavelength:
    491 - 497 nm
  • Fluorescence emission max:
    515 - 523 nm
  • Purity (HPLC):
    ≥90%

Specification Test Results

About this item

 :
 :472.50 g/mol
Melting Pt:≥250 °C (lit.)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Room Temperature
CAS Number:75168-11-5
Formula:C₂₆H₃₈BrN₃
MDL Number:MFCD00467605