Order Entry
Slovenia
ContactUsLinkComponent
8-Hydroxyjulolidine-9-carboxaldehyde 98%, Sigma-Aldrich®
Irritant
8-Hydroxyjulolidine-9-carboxaldehyde 98%, Sigma-Aldrich®
Catalog # 404640-1G
CAS Number:  
Irritant
8-Hydroxyjulolidine-9-carboxaldehyde 98%, Sigma-Aldrich®
Catalog # 404640-1G
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance (Colour): Yellow to Tan to Green:
    Conforms to Requirements
  • Appearance (Form):
    Powder or Crystals
  • Infrared spectrum:
    Conforms to Structure
  • Carbon:
    70.0 - 73.7%
  • Nitrogen:
    6.2 - 6.6%
  • Purity (GC):
    ≥97.5%

Specification Test Results

About this item

 :
 :217.27 g/mol
Melting Pt:72...74 °C (lit.)
 :M-Clarity™ quality level = MQ100
EINECS:263-966-4
 :GHS07
Storage Temperature:Room Temperature
CAS Number:63149-33-7
Formula:C₁₃H₁₅NO₂
MDL Number:MFCD00192477