Order Entry
Slovenia
ContactUsLinkComponent
2-(3-Aminophenylsulphonyl)ethanol hydrochloride 97%, Sigma-Aldrich®
2-(3-Aminophenylsulphonyl)ethanol hydrochloride 97%, Sigma-Aldrich®
Catalog # 361526-1G
CAS Number:  
2-(3-Aminophenylsulphonyl)ethanol hydrochloride 97%, Sigma-Aldrich®
Catalog # 361526-1G
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to brown powder, crystals or crystalline powder
  • Infrared spectrum:
    Conforms to structure
  • Titration:
    96.5 - 103.5% HClO4
  • High pressure liquid:
    96.5% (minimum)

Specification Test Results

About this item

 :
 :237.70 g/mol
 :GHS07
Melting Pt:210 °C (dec., lit.)
Storage Temperature:Room Temperature
 :M-Clarity™ quality level = MQ100
CAS Number:19076-03-0
Formula:H₂NC₆H₄SO₂CH₂CH₂OH·HCl
MDL Number:MFCD00134194