Eosin B
Supplier: Roth Carl
Certificates
About this item
Synonyms: Eosin bluish, 2,7-Dinitro-4,5-dibromofluorescein, Eosin B disodium salt, Sodium 2-(4,5-dibromo-2,7-dinitro-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate, 2-(4,5-dibromo-3,6-dihydroxy-2,7-dinitroxanthen-9-yl)-benzoic acid disodium salt, Acid Red 91
CAS Number:548-24-3
:
Formula:C₂₀H₆Br₂N₂Na₂O₉
MDL Number:MFCD00005041
Molecular Weight:624.06 g/mol
Melting Pt:> 310 °C
Merck Index:12,03638
Storage Temperature:Ambient
EINECS:208-943-1
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together