
Methyl-2-chloro-4-nitrobenzoate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13324-11-3
:
Formula:C₈H₆ClNO₄
MDL Number:MFCD00017013
:215.59 g/mol
Melting Pt:74…78 °C
EINECS:236-362-3
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together