Sodium nitrite 99.99%, HighPurS-CERT
Catalog # ACSDPSNANIC.100G
Supplier: CPACHEM
New Product
CAS Number:
Specifications
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
About this item
Synonyms: Diazotizing salts
:
:69 g/mol
Boiling Pt:600...700 °C
Melting Pt:271 °C
Density:2,168 g/cm³
EINECS:231-555-9
UN:1500
ADR:5.1,III
:GHS03
:GHS06
:GHS09
CAS Number:7632-00-0
Formula:NaNO₂
MDL Number:MFCD00011118