2,5-Dibutoxy-4-(4-morpholinyl)benzenediazonium tetrafluoroborate
Catalog # APOSOR98027
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:421.24 g/mol
CAS Number:50543-78-7
Formula:C₁₈H₂₈BF₄N₃O₃
MDL Number:MFCD00134254