ESD Safety Shoes, Lace-up, Low, Jori jo_Champ Low S3
Supplier: ELTEN
Certificates
About this item
ESD safety shoe made from cowhide leather upper and a PU/PU sole (jo_Explore).
- Fabric lining for natural ventilation
- Full, padded bellows tongue
- Full-length inlay sole jo_TITAN ESD
- Soft fleece ESD insole
- Steel midsole
- Steel toe cap
- PU scuff cap
Suitable for indoors and outdoors, areas where exposure to moisture is expected (S2), areas where there is a risk of penetration from pointed and sharp objects (S3), areas where there is a risk of electrostatic discharge (ESDS/ESD).
Colour: Black
Certifications: EN ISO 20345 S3 FO/LG/SR, form A.
Specifications
- Catalog No:
- ELTN12421/46
- ELTN12421/45
- ELTN12421/47
- ELTN12421/40
- ELTN12421/48
- ELTN12421/41
- ELTN12421/42
- ELTN12421/43
- ELTN12421/36
- ELTN12421/44
- ELTN12421/37
- ELTN12421/38
- ELTN12421/39
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001