Order Entry
Slovenia
ContactUsLinkComponent
Anti-TKFC Rabbit Polyclonal Antibody
Anti-TKFC Rabbit Polyclonal Antibody
Catalog # PRSI16-040
Supplier:  ProSci Inc.
Anti-TKFC Rabbit Polyclonal Antibody
Catalog # PRSI16-040
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Triokinase and FMN cyclase
  • Antigen symbol:
    TKFC
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    26007
  • Antigen synonyms:
    dihydroxyacetone kinase 2 homolog|Dha kinase/FMN cyclase|DHA kinase|NET45|bifunctional ATP-dependent dihydroxyacetone kinase/FAD-AMP lyase (cyclizing)|triokinase/FMN cyclase|DAK|ATP-dependent dihydroxyacetone kinase|FAD-AMP lyase cyclic FMN forming|glycerone kinase|FAD-AMP lyase cyclizing|testis tissue sperm-binding protein Li 84P
  • Modification:
    None
  • Storage buffer:
    PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
  • Storage temperature:
    Store at −20 °C. Avoid freeze/thaw cycles
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 226-575 of human TKFC (NP_056348.2)
  • Purification:
    Affinity purification
  • Pack type:
    Vial
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: TKFC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat