Anti-MCCC2 Rabbit Polyclonal Antibody
Catalog # PRSI15-917
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Methylcrotonoyl-CoA carboxylase beta chain, mitochondrial
- Antigen symbol:MCCC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:64087
- Antigen synonyms:3-methylcrotonyl-CoA:carbon dioxide ligase subunit beta|MCCase subunit beta|MCCC2|MCCB|mitochondrial|3-methylcrotonyl-CoA carboxylase non-biotin-containing subunit|3-methylcrotonyl-CoA carboxylase 2|Methylcrotonoyl-CoA carboxylase beta chain
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 284-563 of human MCCC2 (NP_071415.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: MCCC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat