Anti-COL1A1 Mouse Monoclonal Antibody [clone: 4F6] (Biotinylated)
Catalog # PRSI98-526
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:collagen type I alpha 1 chain
- Antigen symbol:COL1A1
- Clonality:Monoclonal
- Clone:[4F6]
- Conjugation:Biotin
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Western blot:Yes
- Form:Liquid
- Gene ID:1277;1278
- Antigen synonyms:COL1
- Modification:None
- Storage buffer:Supplied in PBS/NaN₃
- Storage temperature:Store vial at 2...8 °C
- Concentration:0,5 mg/ml
- Shipping temperature:20
- Immunogen:Native human type I collagen
- Pack type:Vial
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: COL1A1
Clonality: Monoclonal
Clone: [4F6]
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human