Anti-CRCP Rabbit Polyclonal Antibody
Catalog # PRSI19-435
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:CGRP receptor component
- Antigen symbol:CRCP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:27297
- Antigen synonyms:RNA polymerase III subunit C9|DNA-directed RNA polymerase III subunit RPC9|RCP|CGRP-RCP|calcitonin gene-related peptide-receptor component protein|CGRPRCP|RPC9|CGRP-receptor component protein|RCP9
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-115 of human CRCP (NP_001035737.1).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: CRCP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat