Order Entry
Slovenia
ContactUsLinkComponent
Anti-MAP7 Rabbit Polyclonal Antibody
Anti-MAP7 Rabbit Polyclonal Antibody
Catalog # PRSI19-324
Supplier:  ProSci Inc.
Anti-MAP7 Rabbit Polyclonal Antibody
Catalog # PRSI19-324
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Microtubule associated protein 7
  • Antigen symbol:
    MAP7
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    9053
  • Antigen synonyms:
    MAP-7|dJ325F22.2 (microtubule-associated protein 7 (EMAP115|EMAP115|E-MAP-115))|ensconsin|E-MAP-115|epithelial microtubule-associated protein of 115 kDa
  • Storage buffer:
    PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
  • Storage temperature:
    Store at −20...+730 °C. Avoid freeze/thaw cycles.
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 490-749 of human MAP7 (NP_003971.1).
  • Purification:
    Affinity purification
  • Pack type:
    Vial
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: MAP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse