Anti-SLC13A4 Rabbit Polyclonal Antibody
Catalog # PRSI19-059
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Solute carrier family 13 member 4
- Antigen symbol:SLC13A4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:26266
- Antigen synonyms:solute carrier family 13 member 4|solute carrier family 13 (sodium/sulfate symporter)|member 4|solute carrier family 13 (sodium/sulfate symporters)|Na(+)/sulfate cotransporter SUT-1|NAS2|solute carrier family 13 (sodium/sulphate symporters)|sulphate transporter 1|SUT-1|SUT1
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 390-465 of human SLC13A4 (NP_036582.2).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SLC13A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat