Anti-NR2E3 Rabbit Polyclonal Antibody
PRSI57-558
- Antibody type:Primary
- Antigen name:Photoreceptor-specific nuclear receptor
- Antigen symbol:NR2E3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:170-198 aa
- Form:Liquid
- Gene ID:10002
- Modification:None
- Molecular weight:45 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,36 mg/ml
- Shipping temperature:4 °C
- Immunogen:This NR2E3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 170-198 amino acids from the Central region of human NR2E3
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
For WB starting dilution is: 1:1000
Type: Primary
Antigen: NR2E3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 170-198
Host: Rabbit
Isotype: Ig
Reactivity: Human