Anti-OR4C16 Rabbit Polyclonal Antibody
PRSI57-161
- Antibody type:Primary
- Antigen name:Olfactory receptor 4C16
- Antigen symbol:OR4C16
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:282-310 aa
- Form:Liquid
- Gene ID:219428
- Modification:None
- Molecular weight:35 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,44 mg/ml
- Shipping temperature:4 °C
- Immunogen:This OR4C16 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 282-310 amino acids from the C-terminal region of human OR4C16
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
For WB starting dilution is: 1:1000
Type: Primary
Antigen: OR4C16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 282-310
Host: Rabbit
Isotype: Ig
Reactivity: Human