Anti-ZFPM2 Rabbit Polyclonal Antibody
PRSI58-218
- Antibody type:Primary
- Antigen name:Zinc finger protein ZFPM2
- Antigen symbol:ZFPM2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:883 - 911 aa
- Form:Liquid
- Gene ID:23414
- Modification:None
- Molecular weight:128 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This ZFPM2 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 883-911 amino acids from the C-terminal region of human ZFPM2
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: ZFPM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 883-911
Host: Rabbit
Isotype: Ig
Reactivity: Human