Anti-KIF5C Rabbit Polyclonal Antibody
PRSI57-776
- Antibody type:Primary
- Antigen name:Kinesin heavy chain isoform 5C
- Antigen symbol:KIF5C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Mouse
- Western blot:Yes
- Epitope:758 - 786 aa
- Form:Liquid
- Gene ID:3800
- Modification:None
- Molecular weight:109 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,39 mg/ml
- Shipping temperature:4 °C
- Immunogen:This KIF5C antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 758-786 amino acids from the C-terminal region of human KIF5C
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: KIF5C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 758-786
Host: Rabbit
Isotype: Ig
Reactivity: Mouse