Anti-Recombinant Antibody
Catalog # PRSI10-163
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Clonality:Recombinant
- Conjugation:Unconjugated
- Isotype:F(ab)2 kappa
- Antigen synonyms:F(ab*)2 4D5-8/UCHT1-v1|4D5-8 and UCHT1|BsF(ab')2 v1|3-4D10|F(ab')2 trastusumab/humanized UCHT1-v1|225.28S|Tecnemab-K-1
- Modification:None
- Storage buffer:PBS buffer pH 7,5
- Storage temperature:Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2…8 °C for 1 - 2 weeks. Store at –20 °C for 12 months. Store at –80 °C for long term storage.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Mus musculus,Humanised/CSPG4/HMW-MAA/CD3E/ERBB2 [Homo sapiens],toxin [Clostridium perfringens]
- Purification:>95%
- Pack type:Vial
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: F(ab')2, Fab / Fab2 mix, IgG1-nd, F(ab')2-G1-kappa
Reactivity: