Anti-Spike Protein Recombinant Antibody [clone: 2146] (Biotinylated)
Catalog # PRSI10-056
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Surface glycoprotein
- Antigen symbol:Spike Protein
- Clonality:Recombinant
- Clone:2146
- Conjugation:Biotin
- ELISA:Yes
- ImmunoChemistry:Yes
- Reactivity:Virus
- Form:Liquid
- Antigen synonyms:SARS-CoV2 Spike NTD|Severe acute respiratory syndrome coronavirus 2|SARS-CoV2|2019-nCoV|COVID-19
- Storage buffer:This biotinylated monoclonal antibody is formulated in 0,01 M phosphate buffered saline (PBS) pH 7,4, 1% BSA and 0,09% sodium azide as a preservative
- Storage temperature:This biotinylated monoclonal antibody is stable when stored at 2…8 °C. Do not freeze. Thaw cycles.
- Concentration:0,5 mg/ml
- Shipping temperature:20
- Immunogen:Sequenced from human survivors of COVID-19 (SARS-CoV-2). The spike NTD is expressed on the surface of SARS-CoV-2.
- Pack type:Vial
- Pk:0,05 mg
Specifications
About this item
Type: Primary
Antigen: Spike protein
Clonality: Recombinant
Clone: 2146
Conjugation: biotin
Epitope:
Host:
Isotype: Human IgG1
Reactivity: Virus