Anti-Spike Protein Recombinant Antibody [clone: 2165] (Biotinylated)
PRSI10-053
New Product
- Antibody type:Primary
- Antigen name:Surface glycoprotein
- Antigen symbol:Spike Protein
- Clonality:Recombinant
- Clone:2165
- Conjugation:Biotin
- ELISA:Yes
- ImmunoChemistry:Yes
- Form:Liquid
- Antigen synonyms:Receptor Binding Domain Monoclonal Antibody|SARS-CoV-2 Spike RBD Antibody
- Storage buffer:This biotinylated monoclonal antibody is formulated in 0,01 M phosphate buffered saline (PBS) pH 7,4, 1% BSA and 0,09% sodium azide as a preservative
- Storage temperature:This biotinylated monoclonal antibody is stable when stored at 2...8 °C. Do not freeze.
- Concentration:0,5 mg/ml
- Shipping temperature:20
- Immunogen:Sequenced from human survivors of COVID-19 (SARS-CoV-2). The spike RBD is expressed on the surface of SARS-CoV-2.
- Pack type:Vial
- Pk:0,05 mg
Type: Primary
Antigen: Spike protein
Clonality: Recombinant
Clone: 2165
Conjugation: biotin
Epitope:
Host:
Isotype: Human IgG1
Reactivity: Virus