Anti-CYP19A1 Rabbit Polyclonal Antibody
PRSI18-546
- Antibody type:Primary
- Antigen name:Cytochrome P450, family 19, subfamily A, polypeptide 1
- Antigen symbol:CYP19A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:1588
- Antigen synonyms:ARO1|subfamily A|CYAR|cytochrome P450|microsomal monooxygenase|ARO|subfamily XIX (aromatization of androgens)|P-450AROM|polypeptide 1|CYP19A1|CPV1|MGC104309|aromataseN: cytochrome P450|flavoprotein-linked monooxygenase|CYP19|estrogen synthetase|family 19
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 234-503 of human Aromatase (NP_000094.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: CYP19A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat