Anti-FTH1 Rabbit Polyclonal Antibody
Catalog # PRSI62-861
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Ferritin heavy chain
- Antigen symbol:FTH1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:153-180 aa
- Form:Liquid
- Gene ID:2495
- Modification:None
- Molecular weight:21 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Shipping temperature:4 °C
- Immunogen:This FRIH antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 153-180 amino acids from the C-terminal region of human FRIH.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: FTH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 153-180
Host: Rabbit
Isotype: Ig
Reactivity: Human