Anti-CIB1 Rabbit Polyclonal Antibody
PRSI60-813
- Antibody type:Primary
- Antigen name:Calcium and integrin-binding protein 1
- Antigen symbol:CIB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:122 - 151 aa
- Form:Liquid
- Gene ID:10519
- Modification:None
- Molecular weight:22 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This CIB1 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 122 - 151 amino acids from the C-terminal region of human CIB1.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: CIB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 122-151
Host: Rabbit
Isotype: Ig
Reactivity: Human