Anti-ADRA1B Rabbit Polyclonal Antibody
Catalog # PRSI60-786
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Alpha-1B adrenergic receptor
- Antigen symbol:ADRA1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:380 - 409 aa
- Form:Liquid
- Gene ID:147
- Modification:None
- Molecular weight:57 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This ADRA1B antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 380 - 409 amino acids from the C-terminal region of human ADRA1B.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: ADRA1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 380-409
Host: Rabbit
Isotype: Ig
Reactivity: Human