Anti-VGLL4 Rabbit Polyclonal Antibody
Catalog # PRSI60-565
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Transcription cofactor vestigial-like protein 4
- Antigen symbol:VGLL4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:136 - 163 aa
- Form:Liquid
- Gene ID:9686
- Modification:None
- Molecular weight:31 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,48 mg/ml
- Shipping temperature:4 °C
- Immunogen:This VGLL4 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 136 - 163 amino acids from the Central region of human VGLL4.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: VGLL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 136-163
Host: Rabbit
Isotype: Ig
Reactivity: Human