Anti-DUOX1 Rabbit Polyclonal Antibody
PRSI23-672
New Product
- Antibody type:Primary
- Antigen name:dual oxidase 1
- Antigen symbol:DUOX1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:53905
- Antigen synonyms:THOX1|NADPH thyroid oxidase 1|MGC138841|flavoprotein NADPH oxidase|dual oxidase 1|long NOX 1|NOXEF1|nicotinamide adenine dinucleotide phosphate oxidase|MGC138840|LNOX1|large NOX 1|thyroid oxidase 1|DUOX1
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1312-1551 of human DUOX1 (NP_059130.2)
- Purification:Affinity purification.
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: DUOX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat