Anti-dsRNA Recombinant Antibody [Clone: 1D3]
Catalog # PRSI24-396
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Clonality:Recombinant
- Clone:1D3
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Virus
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:double-stranded RNA|double-stranded ribonucleic acid
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:RDV-RNA-methylated bovine serum albumin complex
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: dsRNA
Clonality: Recombinant
Clone: 1D3
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1 kappa
Reactivity: Virus