Anti-SCARB1 Recombinant Antibody [Clone: 185-1G2 (B467)]
Catalog # PRSI24-751
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:HGNC:HGNC:1664
- Antigen symbol:SCARB1
- Clonality:Recombinant
- Clone:185-1G2 (B467)
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgM kappa
- Reactivity:
- Environmentally Preferable:
- Cross adsorption:No
- Form:Liquid
- Gene ID:949
- Antigen synonyms:SR-BI|CD36 antigen-like 1|Collagen type I receptor|CD36 and LIMPII analogous 1|SRB1|CLA-1|thrombospondin receptor-like 1|Scavenger receptor class B member 1
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunizing mice with stimulated human leukocytes
- Tested applications:Blocking
- Pack type:Vial
- Pk:0,05 mg
Specifications
About this item
Type: Primary
Antigen: HGNC:HGNC:1664
Clonality: Recombinant
Clone: 185-1G2 (B467)
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgM kappa
Reactivity: Human