Trypsin
—, 100 µG
Supplier: ROCHE
Jump to:
Product Details & Documents
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:—
- Pk:100 µG
- Cat. No.:ROCH11418475001
- Storage conditions:Refrigerator
- MDL No.:MFCD00082094
- Enzyme name:Trypsin
- CAS No.:9002-07-7
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



