Order Entry
Slovenia
Orders LinkContactUsLinkComponent
4-(Dimethylamino)benzaldehyde
 :   Roth Carl
4-(Dimethylamino)benzaldehyde
 :  Roth Carl
4-(Dimethylamino)benzaldehyde
4-(Dimethylamino)benzaldehyde
 :  Ehrlich's Reagent, 4-(N,N-Dimethylamino)benzaldehyde, p-Dimethylamino-Benzaldehyde, p-Dimethylaminobenzaldehyde, p-(Dimethylamino)benzaldehyde
  •  :100-10-7
  •  :(CH₃)₂NC₆H₄CHO
  •  :MFCD00003381
  •  :149.19 g/mol
  •  :13,03257
  •  :176…177 °C (1013 hPa)
  •  :72 °C
  •  :1.0254 g/cm³ (99.9 °C)
  •  :>100 °C
  •  :Ambient
  •  :202-819-0
Product Family Options
  • Pack type
    Pk
  • Glass bottle for solids
    25 g
    1 * 25 g
    1 * 25 g - 57,00 €
    57,00 €
    1 * 25 g
       57,00 €
  • Glass bottle for solids
    100 g
    1 * 100 g
    1 * 100 g - 145,00 €
    145,00 €
    1 * 100 g
       145,00 €