Xestospongin C ≥90% (by TLC)
Catalog # ENZOBMLCA4090050
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:50 µG
- Seq:1202
Specifications
About this item
:
:446.71 g/mol
Storage Temperature:Freezer
CAS Number:88903-69-9
Formula:C₂₈H₅₀N₂O₂
MDL Number:MFCD01862629