

Guanfacine hydrochloride ≥99% (by TLC)
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
CAS Number:29110-48-3
:29520-14-7
Formula:C₉H₉Cl₂N₃O
MDL Number:MFCD00798230
:282.55 g/mol
UN:2811
ADR:6.1,III
:GHS06
:GHS08
Specification Test Results
- Purity:≥99% (HPLC)
- Identity:Determined by NMR
Specification Test Results
Frequently Bought Together