
NBQX ≥99% (by HPLC)
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
CAS Number:118876-58-7
:
Formula:C₁₂H₈N₄O₆S
MDL Number:MFCD00270054
:336.28 g/mol
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Purity (HPLC):≥99%
Specification Test Results
Frequently Bought Together