Order Entry
Slovenia
ContactUsLinkComponent
Olomoucine ≥98%, high purity
Olomoucine ≥98%, high purity
  ENZOALX350013M025
 :  
Olomoucine ≥98%, high purity
  ENZOALX350013M025
 :  

 

  • Pk:
    25 mg
  • Seq:
    2315

 

 

 :
 :298.343 g/mol
 :101622-51-9
 :C₁₅H₁₈N₆O
 :MFCD00189360