H-1152 dihydrochloride ≥98% (by HPLC)
:45146-2-58-1
:
:C₁₆H₂₁N₃O₂S·2HCl
:319.4 · 73.0 g/mol
:Freezer
Frequently Bought Together
Transfer pipettes
: RATIOLAB
Reagent/centrifuge tube with screw cap
: Greiner Bio-One
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
VWR®, Petri Dish, PS
: VWR Collection
VWR®, Centrifuge Tubes, High Performance, Flat or Plug Cap, PP
: VWR Collection
VWR®, Cryoboxes, Cardboard, 136x136 mm
: VWR Collection
Cell culture flasks, Nunc™
: Thermo Fisher Scientific
VWR®, Cryovials
: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
Petri dishes
: Greiner Bio-One
VWR®, Spreaders, L-shaped, Disposable
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection