Order Entry
Slovenia
ContactUsLinkComponent
7-HC-6-heptenoate ≥98% (by TLC)
Irritant
7-HC-6-heptenoate ≥98% (by TLC)
Catalog # ENZOBMLST5010010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Irritant
7-HC-6-heptenoate ≥98% (by TLC)
Catalog # ENZOBMLST5010010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

Specification Test Results

  • Purity:
    ≥98% (TLC)

Specification Test Results

About this item

 :
 :272.3 g/mol
 :GHS07
Storage Temperature:Ambient
Formula:C₁₆H₁₆O₄