NNGH ≥99% (by TLC)
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
CAS Number:161314-17-6
:
MDL Number:MFCD02179370
Frequently Bought Together
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Sampling Spoons
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
VWR®, Transfer Pipettes, Graduated
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR® Blender Bags
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Cryoboxes with Dividers
Supplier: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
VWR®, Petri Dish, PS
Supplier: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF