AH-6809 ≥98% (by TLC)
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
CAS Number:33458-93-4
:
Formula:C₁₇H₁₄O₅
MDL Number:MFCD08056083
:298.3 g/mol
Storage Temperature:Ambient
Specification Test Results
- Purity:≥98% (TLC)
Specification Test Results
Frequently Bought Together