

Gambogic amide ≥98% (by TLC)
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
CAS Number:286935-60-2
:
Formula:C₃₈H₄₅NO₇
:627.8 g/mol
Melting Pt:74…76 °C
:GHS06
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Purity:≥98% (TLC)
- Appearance:Yellow-orange solid
- Melting Point:74 - 76°C
Specification Test Results
Frequently Bought Together