Order Entry
Slovenia
ContactUsLinkComponent
DL-Thiorphan ≥97% (by TLC)
DL-Thiorphan ≥97% (by TLC)
Catalog # ENZOBMLPI1130005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
DL-Thiorphan ≥97% (by TLC)
Catalog # ENZOBMLPI1130005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :253.316 g/mol
Storage Temperature:Freezer
CAS Number:76721-89-6
Formula:C₁₂H₁₅NO₃S
MDL Number:MFCD00058435