Order Entry
Slovenia
ContactUsLinkComponent
Ethidium bromide Greenbag™ disposal kit, MP Biomedicals
Ethidium bromide Greenbag™ disposal kit, MP Biomedicals
Catalog # ICNA112350200
Supplier:  MP Biomedicals
CAS Number:  
Ethidium bromide Greenbag™ disposal kit, MP Biomedicals
Catalog # ICNA112350200
Supplier:  MP Biomedicals
CAS Number:  
Specifications
  • Pk:
    50
  • Description:
    Ethidium Bromide Greenbag™ Disposal Kit
Specifications
About this item
The EtBr Greenbag™ Disposal Kit allows rapid and trouble-free concentration of ethdium bromide from large volumes of solutions into a small 'teabag' containing activated carbon. The Greenbag™ is then conveniently disposed of along with other solid hazardous wastes.
One kit has the capacity to remove 500 mg of ethidium bromide from solutions (10 mg EtBr/bag), enough to satisfy the requirements of most laboratories for a year.