Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Phosphoryl trichloride, Msynth® plus, Sigma-Aldrich®
Phosphoryl trichloride, Msynth® plus, Sigma-Aldrich®
 :  
 :  10025-87-3
Phosphoryl trichloride, Msynth® plus, Sigma-Aldrich®
 :  10025-87-3
 :  Phosphorus (V) oxide chloride, Phosphorus oxychloride, Phosphoryl chloride, Phosphorus(V) oxychloride
  •  :10025-87-3
  •  :POCl₃
  •  :MFCD00011443
  •  :153.33 g/mol
  •  :12,07506
  •  :107 °C (1013 hPa)
  •  :1 °C
  •  :1.645 g/cm³ (20 °C)
  •  :Ambient
  •  :233-046-7
  •  :6.1 (8),I
  •  :M-Clarity™ quality level = MQ400
  •  :1810