Order Entry
Slovenia
ContactUsLinkComponent
Safety Shoes, Lace-Up, S3 Spes II
Safety Shoes, Lace-Up, S3 Spes II
Catalog # ENGE93924
Supplier:  ENGELHARDT
Safety Shoes, Lace-Up, S3 Spes II
Catalog # ENGE93924
Supplier:  ENGELHARDT
Specifications
  • Shoe size:
    43
  • Colour:
    Black
  • Pk:
    1 PAIR
Specifications
About this item
Premium leather, high-performance membrane and the full S3 protection package.
  • Durable safety shoes in a sneaker style
  • Waterproof, windproof and breathable thanks to the dryplexx® membrane
  • Outer material in high-quality, full-grain calf's leather
  • Genuine leather
  • Breathable mesh inner lining
  • Dirt cannot penetrate thanks to the closed tongue design
  • Removable, full-surface anatomically shaped insole
  • Anti-slip rubber sole conforms to SRA, anti-static, fuel-resistant and heat-resistant up to approx. 180 °C
The very casual style of the S3 safety shoes Spes stands out, especially as far as weather protection and durability is concerned. The breathable mesh lining combined with the dryplexx® membrane technology also guarantees a pleasant climate inside the shoes at all times - even on warm days.
Sole material: Rubber
Certifications: EN ISO 20345:2011 S3 with steel toe cap and steel midsole - the properties are similar to those of the new EN ISO 20345:2022 S7.