Custom Volatile Mixture
Catalog # EHERQ60014672
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pk:1
- Description:Custom volatile mixture 14672 1000 µg/ml in methanol
- UN No.:1230
- GHS signal:Danger
- Melting point:−98 °C
- Colour:Colourless
- Flash point:11 °C
- Odour/scent:Alocohol
- Boiling point:64,72 °C
- Form:Liquid
Specifications
About this item
Caution: Laboratory use only.