Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Custom Pesticide Mix
Custom Pesticide Mix
Custom Pesticide Mix
Catalog # EHERQ60014670
Supplier:
EHRENSTORFER
CAS Number:
Expand image
Custom Pesticide Mix
Catalog # EHERQ60014670
Supplier:
EHRENSTORFER
CAS Number:
Specifications
Pk:
1
Description:
Custom pesticide mix 14670 100 mg/L in acetone
UN No.:
1090
GHS signal:
Warning
Specifications
About this item
Product Details
Product Details