Order Entry
Slovenia
ContactUsLinkComponent
Baktolan® Lotion
Baktolan® Lotion
Catalog # BODL980171
Supplier:  Bode
Baktolan® Lotion
Catalog # BODL980171
Supplier:  Bode

Specifications

  • Description:
    Lotion
  • Pk:
    20

Specifications

About this item

Moisturising, fast-absorbing fragrance-free oil-in-water (o/w) lotion for normal skin.

  • Fragrance-free
  • Nurturing formula with plant-based oils and vitamin E
  • Increases significantly the skin's moisture content is quickly absorbed leaving no residues
  • Provides a smooth and soft skin feeling
  • Free of parabens
  • Ideal for the quick skin care in between
  • Appearance: White to slightly white emulsion
  • Density: 0,990 g/cm³
  • pH value: 5,5 to 6,5

Baktolan lotion pure is a fragrance-free light oil-in-water lotion matched to the skin's pH-value. Particular value was placed on very good skin compatibility and quick absorption when using the lotion. The nurturing formula with plant-based oils and vitamin E provides a perceptibly smooth and soft feeling of the skin.

Certifications: EN 455-1, EN455-2, EN1500.